Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499672.2 | XP_003499720.1 | 1 - 2 | 1917 | 71 | |
XM_007640395.1 | XP_007638585.1 | 1 - 2 | 1917 | 71 | ||
XM_007640396.1 | XP_007638586.1 | 1 - 2 | 1917 | 71 | ||
XM_007640397.1 | XP_007638587.1 | 1 - 2 | 1917 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |