Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503890.2 | XP_003503938.1 | 3 - 4 | 457 | 99 | |
GenBank mRNA | JI879363.1 | - | 3 - 4 | 449 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |