Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510195.1 | XP_003510243.1 | 3 - 4 | 477 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Protease Probe spans exons |