Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sys1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645123.1 | XP_007643313.1 | 3 - 4 | 661 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |