Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cript | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511401.2 | XP_003511449.1 | 2 - 3 | 152 | 90 | |
GenBank mRNA | JI889249.1 | - | 2 - 3 | 145 | 90 | |
JP058223.1 | - | 2 - 3 | 145 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |