Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647389.1 | XP_007645579.1 | 3 - 4 | 439 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Protease Detect Genomic DNA Recommended Assay Probe spans exons |