Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503244.2 | XP_003503292.1 | 2 - 3 | 1160 | 88 | |
XM_007643848.1 | XP_007642038.1 | 2 - 3 | 1165 | 88 | ||
GenBank mRNA | JP044056.1 | - | 1 - 2 | 152 | 88 |