Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znf76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506290.2 | XP_003506338.1 | 1 - 2 | 175 | 68 | |
XM_007646796.1 | XP_007644986.1 | 1 - 2 | 176 | 68 | ||
GenBank mRNA | JI877658.1 | - | 1 - 2 | 100 | 68 | |
JP051495.1 | - | 1 - 2 | 100 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |