Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509370.1 | XP_003509418.1 | 3 - 4 | 1050 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |