Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507283.2 | XP_003507331.1 | 3 - 4 | 350 | 102 | |
GenBank mRNA | JP061642.1 | - | 3 - 4 | 313 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Probe spans exons |