Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scxb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507665.2 | XP_003507713.1 | 1 - 2 | 2074 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |