Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcamr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007616739.1 | XP_007614929.1 | 4 - 5 | 1246 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |