Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100754573 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654419.1 | XP_007652609.1 | 6 - 7 | 2221 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |