Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kazald1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512837.2 | XP_003512885.1 | 3 - 4 | 1366 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |