Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptplb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640866.1 | XP_007639056.1 | 2 - 3 | 240 | 75 | |
GenBank mRNA | JI884823.1 | - | 2 - 3 | 240 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Protein Phosphatase Recommended Assay Probe spans exons |