Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il22ra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496558.1 | XP_003496606.1 | 1 - 2 | 57 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |