Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lysmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498665.2 | XP_003498713.2 | 2 - 3 | 323 | 77 | |
GenBank mRNA | JI867761.1 | - | 1 - 2 | 194 | 77 | |
JP064660.1 | - | 1 - 2 | 194 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons |