Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lysmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498665.2 | XP_003498713.2 | 3 - 4 | 686 | 86 | |
GenBank mRNA | JI867761.1 | - | 2 - 3 | 557 | 86 | |
JP064660.1 | - | 2 - 3 | 557 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |