Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qtnf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498815.2 | XP_003498863.1 | 2 - 3 | 225 | 97 | |
XM_007633696.1 | XP_007631886.1 | 4 - 5 | 425 | 97 | ||
XM_007633712.1 | XP_007631902.1 | 2 - 3 | 324 | 97 |