Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aven | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503593.2 | XP_003503641.1 | 3 - 4 | 364 | 66 | |
GenBank mRNA | JI892302.1 | - | 3 - 4 | 353 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis Probe spans exons |