Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495268.2 | XP_003495316.1 | 1 | 325 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |