Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640839.1 | XP_007639029.1 | 4 - 5 | 692 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |