Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qtrtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497882.2 | XP_003497930.1 | 9 - 10 | 1373 | 117 | |
XM_007618907.1 | XP_007617097.1 | 9 - 10 | 1490 | 117 | ||
XM_007618916.1 | XP_007617106.1 | 9 - 10 | 1497 | 117 | ||
XM_007618931.1 | XP_007617121.1 | 7 - 8 | 1086 | 117 | ||
GenBank mRNA | JI895502.1 | - | 3 - 4 | 393 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |