Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mff | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514903.2 | XP_003514951.1 | 5 - 6 | 742 | 73 | |
XM_007654938.1 | XP_007653128.1 | 5 - 6 | 742 | 73 | ||
XM_007654940.1 | XP_007653130.1 | 5 - 6 | 742 | 73 | ||
XM_007654941.1 | XP_007653131.1 | 4 - 5 | 567 | 73 | ||
XM_007654942.1 | XP_007653132.1 | 6 - 7 | 815 | 73 | ||
XM_007654943.1 | XP_007653133.1 | 5 - 6 | 640 | 73 | ||
XM_007654944.1 | XP_007653134.1 | 5 - 6 | 742 | 73 | ||
XM_007654945.1 | XP_007653135.1 | 5 - 6 | 742 | 73 | ||
XM_007654946.1 | XP_007653136.1 | 5 - 6 | 742 | 73 | ||
XM_007654947.1 | XP_007653137.1 | 5 - 6 | 742 | 73 | ||
GenBank mRNA | JI862216.1 | - | 3 - 4 | 506 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons |