Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495881.2 | XP_003495929.2 | 3 - 4 | 513 | 95 | |
XM_007644918.1 | XP_007643108.1 | 3 - 4 | 376 | 95 | ||
GenBank mRNA | JI877044.1 | - | 1 - 2 | 186 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |