Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tti2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497839.2 | XP_003497887.1 | 6 - 7 | 1807 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |