Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643426.1 | XP_007641616.1 | 3 - 4 | 460 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |