Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648026.1 | XP_007646216.1 | 2 - 3 | 293 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |