Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647267.1 | XP_007645457.1 | 5 - 6 | 476 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel Probe spans exons |