Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aprt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495090.2 | XP_003495138.1 | 1 - 2 | 158 | 92 | |
GenBank mRNA | JI882185.1 | - | 1 - 2 | 141 | 92 | |
JP061249.1 | - | 1 - 2 | 141 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |