Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aph1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499338.2 | XP_003499386.1 | 3 - 4 | 358 | 60 | |
GenBank mRNA | JI889120.1 | - | 3 - 4 | 482 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |