Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505308.2 | XP_003505356.1 | 3 | 1441 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Ion Channel 3' Most Detect Genomic DNA Both primers and probe map within a single exon |