Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007613857.1 | XP_007612047.1 | 1 - 2 | 73 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |