Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641718.1 | XP_007639908.1 | 2 - 3 | 409 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |