Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502119.1 | XP_003502167.1 | 1 | 324 | 88 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Both primers and probe map within a single exon |