Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyhr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507635.2 | XP_003507683.1 | 1 - 2 | 535 | 86 | |
XM_007648226.1 | XP_007646416.1 | 2 - 3 | 702 | 86 | ||
XM_007648227.1 | XP_007646417.1 | 2 - 3 | 512 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |