Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3galtl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007652246.1 | XP_007650436.1 | 11 - 12 | 1014 | 98 | |
GenBank mRNA | JP038488.1 | - | 11 - 12 | 1114 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |