Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507652.2 | XP_003507700.1 | 4 - 5 | 1268 | 96 | |
XM_007648223.1 | XP_007646413.1 | 4 - 5 | 1262 | 96 | ||
XM_007648225.1 | XP_007646415.1 | 5 - 6 | 1079 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |