Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd11b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007649190.1 | XP_007647380.1 | 1 - 2 | 263 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |