Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pard6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643103.1 | XP_007641293.1 | 2 - 3 | 133 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |