Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495343.2 | XP_003495391.1 | 3 - 4 | 493 | 71 | |
XM_007634199.1 | XP_007632389.1 | 2 - 3 | 525 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |