Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tac4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503964.2 | XP_003504012.2 | 2 - 3 | 961 | 79 | |
XM_007644480.1 | XP_007642670.1 | 2 - 3 | 961 | 79 | ||
XM_007644481.1 | XP_007642671.1 | 2 - 3 | 961 | 79 |