Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cir1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495963.2 | XP_003496011.1 | 7 - 8 | 638 | 92 | |
GenBank mRNA | JI883167.1 | - | 7 - 8 | 551 | 92 | |
JP041612.1 | - | 7 - 8 | 551 | 92 |