Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499551.2 | XP_003499599.1 | 1 - 2 | 551 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Inventoried Probe spans exons |