Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgol2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642652.1 | XP_007640842.1 | 7 - 8 | 3587 | 99 | |
XM_007642653.1 | XP_007640843.1 | 7 - 8 | 3483 | 99 | ||
GenBank mRNA | JI884768.1 | - | 6 - 7 | 3322 | 99 | |
JP046864.1 | - | 6 - 7 | 3319 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |