Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500041.2 | XP_003500089.2 | 1 - 2 | 296 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Probe spans exons |