Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pter | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505037.2 | XP_003505085.1 | 3 - 4 | 783 | 90 | |
XM_007645542.1 | XP_007643732.1 | 2 - 3 | 516 | 90 | ||
XM_007645543.1 | XP_007643733.1 | 4 - 5 | 868 | 90 | ||
XM_007645544.1 | XP_007643734.1 | 3 - 4 | 618 | 90 | ||
XM_007645545.1 | XP_007643735.1 | 2 - 3 | 516 | 90 | ||
XM_007645546.1 | XP_007643736.1 | 2 - 3 | 516 | 90 | ||
XM_007645547.1 | XP_007643737.1 | 2 - 3 | 516 | 90 | ||
GenBank mRNA | JI878796.1 | - | 1 - 2 | 460 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |