Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfral | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495051.1 | XP_003495099.1 | 5 - 6 | 695 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |