Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640957.1 | XP_007639147.1 | 2 - 3 | 279 | 94 | |
GenBank mRNA | JP040944.1 | - | 2 - 3 | 289 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |