Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olig3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496546.1 | XP_003496594.1 | 1 | 414 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |